Learn More
Sklar Instruments Surgi-OR TC Derf Needle Holder, Sklar 4-3/4 in., Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA.
SDP
Catalog No. 501184170 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR TC Derf Needle Holder, Sklar. 4-3/4 in., Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Surgi-OR TC Derf Needle Holders feature short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures. Frequently utilized in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf Needle Holder to be easily controlled in smaller surgical areas. Hand-crafted in Pakistan, Surgi-OR TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips and a length of 4-3/4 inches.
Catalog No. 50-118-4170 Supplier Sklar Instruments Supplier No. 95136
Surgi-OR TC Derf Needle Holder, Sklar. 4-3/4 in., Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Surgi-OR TC Derf Needle Holders feature short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures. Frequently utilized in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf Needle Holder to be easily controlled in smaller surgical areas. Hand-crafted in Pakistan, Surgi-OR TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips and a length of 4-3/4 inches.