Learn More
Sino Biological DERF1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 69028T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Dermatophagoides farinae DERF1 ( Catalog#69028-A08H; BAC53948.1; Met1-Met321). DERF1 specific IgG was purified by Dermatophagoides farinae DERF1 affinity chromatography.
Catalog No. 50-237-1314