Learn More
Bachem Fmoc-N-Me-Arg(Pbf)-OH
SDP
Supplier:  Bachem 4141352.2002
Encompass_Preferred
Fmoc-N-Me-Arg(Pbf)-OH, 913733-27-4, C35H42N4O7S, Mr 662.81, 5g. Derivative for synthesizing peptides containing Na-methylated arginine, superior to Fmoc-N-Me-Arg(Mtr)-OH (4026642), as Pbf can be removed under much milder conditions.
Catalog No. 50-194-7116