Learn More
Bachem Fmoc-N-Me-Arg(Pbf)-OH
SDP
Catalog No. 501947115
Encompass_Preferred
Fmoc-N-Me-Arg(Pbf)-OH, 913733-27-4, C35H42N4O7S, Mr 662.81, 1g. Derivative for synthesizing peptides containing Na-methylated arginine, superior to Fmoc-N-Me-Arg(Mtr)-OH (4026642), as Pbf can be removed under much milder conditions.
Catalog No. 50-194-7115 Supplier Bachem Supplier No. 4141352.2001
Fmoc-N-Me-Arg(Pbf)-OH, 913733-27-4, C35H42N4O7S, Mr 662.81, 1g. Derivative for synthesizing peptides containing Na-methylated arginine, superior to Fmoc-N-Me-Arg(Mtr)-OH (4026642), as Pbf can be removed under much milder conditions.