Learn More
Bachem Fmoc-(Dmb)Gly-OH
SDP
Supplier:  Bachem 4070304.5000
Encompass_Preferred
Fmoc-(Dmb)Gly-OH, 166881-42-1, C26H25NO6, Mr 447.49, 5g. Fmoc-N-(2,4-dimethoxybenzyl)-Gly-OH. Derivative for the Fmoc-SPPS of backbone-protected peptides. Introduction of Dmb at appropriate positions prevents aggregation and aspartimide formation. Please see also our Dmb-dipeptides.
Catalog No. 50-194-7187