Learn More
Bachem Fmoc-(Dmb)Ala-OH
SDP
Catalog No. 501947088
Encompass_Preferred
Fmoc-(Dmb)Ala-OH, 1425938-66-4, C27H27NO6, Mr 461.51, 1g. Fmoc-N-(2,4-dimethoxybenzyl)-Ala-OH. Derivative for the Fmoc-SPPS of backbone-protected peptides. Introduction of Dmb at appropriate positions prevents aggregation of the growing peptide chain. Conroy et al. could stabilize the aspartimide-prone Asp(OAll or ODmab)-Ala motif by incorporating DmbAla. Please see also our Dmb-dipeptides.
Catalog No. 50-194-7088 Supplier Bachem Supplier No. 4072316.1000
Fmoc-(Dmb)Ala-OH, 1425938-66-4, C27H27NO6, Mr 461.51, 1g. Fmoc-N-(2,4-dimethoxybenzyl)-Ala-OH. Derivative for the Fmoc-SPPS of backbone-protected peptides. Introduction of Dmb at appropriate positions prevents aggregation of the growing peptide chain. Conroy et al. could stabilize the aspartimide-prone Asp(OAll or ODmab)-Ala motif by incorporating DmbAla. Please see also our Dmb-dipeptides.