Learn More
Bachem Fmoc-4-sulfomethyl-Phe(Tce)-OH
SDP
Supplier:  Bachem 4070723.0001
Encompass_Preferred
Fmoc-4-sulfomethyl-Phe(Tce)-OH, 1146758-11-3, C27H24Cl3NO7S, Mr 612.91, 1g. Derivative of a stable sulfotyrosine mimic suitable for Fmoc-SPPS. The lateral trichloroethyl (Tce) group is cleaved by hydrogenolysis (H2/Pd in the presence of ammonium formate).
Catalog No. 50-194-7263