Learn More
Aviva Systems Biology Corp DERL3 Antibody - middle region (ARP44442_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44442_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DERL3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human DERL3. Alternate names: C22orf14, IZP6, LLN2, MGC71803, derlin-3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58809