Learn More
Electron Microscopy Sciences Tripod Polisher Derlin Micrometer Fee
SDP
Catalog No. 50980866
Encompass
The Delrin Feet are applied to the tip of each micrometer and are designed to protect the micrometer as well as the lapping from damage when polishing TEM samples. For use with Tripod Polisher Model 590.
Catalog No. 50-980-866 Supplier Electron Microscopy Sciences Supplier No. 5011615
The Delrin Feet are applied to the tip of each micrometer and are designed to protect the micrometer as well as the lapping from damage when polishing TEM samples. For use with Tripod Polisher Model 590.