Learn More
McKesson General Medical Skin Adhesive McKesson LiquiBand Exceed™ 0.8 mL Liquid Precision and Dome Applicator Tip 2-Octyl Cyanoacrylate
Supplier:  McKesson General Medical 948660
Encompass_Preferred
McKesson LiquiBand Exceed™ Topical Skin Adhesive. For the closure of topical skin incisions including laparoscopic incisions, and trauma-induced lacerations in areas of low skin tension that are simple, thoroughly cleansed, and have easily approximated edges. Featureing impressive yield, 0.8 mL applicator allows for closure of wounds measuring up to 30 cm in length. Ideal for medium to long surgical incisions. No-clog formulation and tip facilitate simple and precise dosage and allow users to work at their own pace with up to 90 minutes of intraoperative use. Elliptical-shaped applicator tip allows for narrow or wide application providing versatility in application technique. Unique tip design provides even, consistent coat, ensures coverage, and prevents stuttering, squirting, and dripping. Pure octyl formulation features increased viscosity for optimal durability and flexibility and creates intimate skin contact.
Catalog No. NC1795873