Learn More
Key Scientific Products Dermapak 2000 - Blue, Compact Design, Lightweight, 100 Per Pack
SDP
Catalog No. NC1232568
Encompass_Preferred
The DERMAPAK 2000 - BLUE is a compact dermatological sample storage solution designed to securely transport skin scrapings and hair samples. Its absorbent properties help reduce bacterial overgrowth during shipping, making it reliable for clinicians and laboratories. Lightweight and cost-effective, the system features a robust design for enhanced convenience and security, distinguishing it from other storage solutions. Ideal for clinical and laboratory environments, it comes in a pack of 100.
Catalog No. NC1232568 Supplier Key Scientific Products Supplier No. DMK202
The DERMAPAK 2000 - BLUE is a compact dermatological sample storage solution designed to securely transport skin scrapings and hair samples. Its absorbent properties help reduce bacterial overgrowth during shipping, making it reliable for clinicians and laboratories. Lightweight and cost-effective, the system features a robust design for enhanced convenience and security, distinguishing it from other storage solutions. Ideal for clinical and laboratory environments, it comes in a pack of 100.