Learn More
Med Vet International Derma Strength Pro, 90 ct Chews
Catalog No. 502828183
Encompass_Preferred
Derma Strength Pro is a hypoallergenic formula featuring essential fatty acids, antioxidants and bioavailable nutrients to support healthy skin structure, texture and resiliency. It is recommended for all dogs and cats in need of additional dermatological support due to allergies, surgeries and other skin afflictions. Active ingredients per chew include: Quercevita™ (Quercetin Phytosome™), 125 mg Omega 6 Fatty Acids, 65 mg Perilla (Perilla frutescens) Seed Extract, 40 mg Omega 3 Fatty Acids, 26 mg Hyaluronic Acid (HyMax Low Molecular Weight Sodium Hyaluronate), 5 mg Oligonol (low-molecular weight Lychee and Green tea extract), 5 mg
Catalog No. 50-282-8183 Supplier Med Vet International Supplier No. PHVDERMASTR90
Derma Strength Pro is a hypoallergenic formula featuring essential fatty acids, antioxidants and bioavailable nutrients to support healthy skin structure, texture and resiliency. It is recommended for all dogs and cats in need of additional dermatological support due to allergies, surgeries and other skin afflictions. Active ingredients per chew include: Quercevita™ (Quercetin Phytosome™), 125 mg Omega 6 Fatty Acids, 65 mg Perilla (Perilla frutescens) Seed Extract, 40 mg Omega 3 Fatty Acids, 26 mg Hyaluronic Acid (HyMax Low Molecular Weight Sodium Hyaluronate), 5 mg Oligonol (low-molecular weight Lychee and Green tea extract), 5 mg