Learn More
ATCC Dermal Cell Basal Medium
Supplier:  ATCC PCS200030
Encompass
Dermal Cell Basal Medium is a sterile, phenol-red-free, liquid tissue culture medium intended for use as one component in a complete ATCC Primary Cell Solutions system. This system is designed to support cells (eg., keratinocytes, melanocytes) derived from a normal human epidermis. Dermal Cell Basal Medium contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts.
Catalog No. 50-238-4751