Learn More
ATCC Dermal Fibroblasts; Normal, Human, Neonatal
Catalog No. 502382857
Encompass
Primary Dermal Fibroblast Normal; Human, Neonatal (HDFn) is a cell line isolated from neonatal foreskin that has research applications in responding to pathogens, skin aging, wound healing, gene delivery, and skin diseases, including scleroderma.
Catalog No. 50-238-2857 Supplier ATCC Supplier No. PCS201010
Primary Dermal Fibroblast Normal; Human, Neonatal (HDFn) is a cell line isolated from neonatal foreskin that has research applications in responding to pathogens, skin aging, wound healing, gene delivery, and skin diseases, including scleroderma.