Learn More
CPC Scientific H-Ala-Leu-Trp-Lys-Thr-Met-Leu-Lys-Lys-Leu-Gly-Thr-Met-Ala-Leu-His-Ala-Gly-Lys-Ala-Ala-Leu-Gly-Ala-Ala-Ala-Asp-Thr-Ile-Ser-Gln-Gly-Thr-Gln-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ATMP005B
Encompass
Peptide that is isolated from skin extracts of the South American arboreal frog, Phyllomedusa Sauvagei. It is a strongly basic, water-soluble, thermalstable nonhemolytic compound, nonhemolytic peptide that exhibits highly potent antimicrobial activity against pathogenic fungi at micromolar concentration. A potential amphiphatic alpha-helix structure can probably function by membrane permeabilization. ONE-LETTER SEQUENCE: ALWKTMLKKLGTMALHAGKAALGAAADTISQGTQMOLECULAR FORMULA: C152H257N43O44S2MOLECULAR WEIGHT:3455.11STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [136033-70-0], RESEARCH AREA: AntimicrobialREFERENCES: A. Mor et al., Biochemistry, 30, 8824 (1991)
Catalog No. 50-210-8817