Learn More
Medchemexpress LLC Dermatan sulphate sodium | 54328-33-5 | 100 MG
SDP
Catalog No. 5000417445
Encompass_Preferred
Dermatan sulphate sodium is an orally active glycosaminoglycan and thrombin inactivator with antithrombotic activity. It selectively catalyzes the heparin cofactor II-mediated inactivation of thrombin without interacting with antithrombin III. This compound promotes stem cell differentiation, exhibits high bioavailability, and alleviates pulmonary fibrosis injury induced by Bleomycin.
  • Thrombin inactivator with antithrombotic activity
  • Selectively catalyzes heparin cofactor II-mediated inactivation of thrombin
  • Promotes stem cell differentiation and enhances neurite outgrowth
  • Exhibits high bioavailability
  • Alleviates pulmonary fibrosis injury and attenuates lung inflammation
  • Useful in research for cardiovascular diseases, inflammation, and stem cell differentiation
Catalog No. 50-004-17445 Supplier Medchemexpress LLC Supplier No. HYN8278100MG
Dermatan sulphate sodium is an orally active glycosaminoglycan and thrombin inactivator with antithrombotic activity. It selectively catalyzes the heparin cofactor II-mediated inactivation of thrombin without interacting with antithrombin III. This compound promotes stem cell differentiation, exhibits high bioavailability, and alleviates pulmonary fibrosis injury induced by Bleomycin.
  • Thrombin inactivator with antithrombotic activity
  • Selectively catalyzes heparin cofactor II-mediated inactivation of thrombin
  • Promotes stem cell differentiation and enhances neurite outgrowth
  • Exhibits high bioavailability
  • Alleviates pulmonary fibrosis injury and attenuates lung inflammation
  • Useful in research for cardiovascular diseases, inflammation, and stem cell differentiation