Learn More
Medchemexpress LLC Dermatan sulphate sodium | 54328-33-5 | 25 MG
SDP
Catalog No. 5000420566
Encompass_Preferred
Dermatan sulphate sodium is an oral active glycosaminoglycan and thrombin inactivator with antithrombotic activity. It selectively catalyzes the heparin cofactor II-mediated inactivation of thrombin without interacting with antithrombin III. Dermatan sulphate sodium promotes stem cell differentiation, exhibits high bioavailability, and alleviates pulmonary fibrosis injury induced by Bleomycin (HY-108345).
  • Oral active glycosaminoglycan and thrombin inactivator
  • Antithrombotic activity
  • Selectively catalyzes heparin cofactor II-mediated inactivation of thrombin
  • Promotes stem cell differentiation
  • Exhibits high bioavailability
  • Alleviates pulmonary fibrosis injury
  • Used in research related to cardiovascular diseases, inflammation, and stem cell differentiation
Catalog No. 50-004-20566 Supplier Medchemexpress LLC Supplier No. 25MGHYN8278
Dermatan sulphate sodium is an oral active glycosaminoglycan and thrombin inactivator with antithrombotic activity. It selectively catalyzes the heparin cofactor II-mediated inactivation of thrombin without interacting with antithrombin III. Dermatan sulphate sodium promotes stem cell differentiation, exhibits high bioavailability, and alleviates pulmonary fibrosis injury induced by Bleomycin (HY-108345).
  • Oral active glycosaminoglycan and thrombin inactivator
  • Antithrombotic activity
  • Selectively catalyzes heparin cofactor II-mediated inactivation of thrombin
  • Promotes stem cell differentiation
  • Exhibits high bioavailability
  • Alleviates pulmonary fibrosis injury
  • Used in research related to cardiovascular diseases, inflammation, and stem cell differentiation