Learn More
Med Vet International Dermatonin (Melatonin) Implant 8mg For Dogs Under 25Lbs
Catalog No. 502828750
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Atypical Cushing's Disease affects a number of dogs and causes hair loss and other problems. Dermatonin (the melatonin implant) for dogs is now available. Veterinary dermatologists are currently engaged in clinical trials using constant-release melatonin implants for the treatment of Atypical Cushing's Disease as well as several other conditions. Dermatonin gives the veterinary market a 100% natural way to treat this condition. There are 3 different sizes available depending on the dog's weight as listed. The implants only need to be administered every 4 to 6 months.
Catalog No. 50-282-8750 Supplier Med Vet International Supplier No. RXDERMATON8
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Atypical Cushing's Disease affects a number of dogs and causes hair loss and other problems. Dermatonin (the melatonin implant) for dogs is now available. Veterinary dermatologists are currently engaged in clinical trials using constant-release melatonin implants for the treatment of Atypical Cushing's Disease as well as several other conditions. Dermatonin gives the veterinary market a 100% natural way to treat this condition. There are 3 different sizes available depending on the dog's weight as listed. The implants only need to be administered every 4 to 6 months.