Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Dermatophyte Milk Agar Tube
SDP

Catalog No. 23001798 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Fill Volume:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
23-001-798 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-798 Supplier Hardy Diagnostics™ Supplier No. L75
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dermatophyte Milk Agar is recommended for use in the cultivation and differentiation of certain dermatophytes, particularly Trichophyton rubrum , Trichophyton mentagrophytes , and Microsporum persicolor

  • Hardy Diagnostics Dermatophyte Milk Agar is prepared according the the formulation of Bromcresol purple-milk solids-glucose medium
  • Alkalinization of Dermatophyte Milk Agar is noted by a color change in the medium from its original blue-gray appearance to a violet purple color
  • Acidification of the medium, noted by a color change from blue-gray to yellow, usually indicates bacterial or yeast contamination

Specifications

Appearance Opaque
Certifications/Compliance ANAB Calibration ISO 17025, BSI-ISO 13485
Color Blue-gray
Content And Storage 2°C to 8°C
Detects Dermatophytes, Trichophyton rubrum, Trichophyton mentagrophytes, Microsporum persicolor, Citrate utilization of gram-negative Enteric bacilli
Format Tube
Manufacturing Origin United States
Media Type Dermatophyte Milk Agar
Product Type Agar Tube
Target Organism Class Fungi
Test Type Microbial Differentiation
Fill Volume 6 mL
For Use With (Application) Cultivation and Differentiation of Dermatophytes, Particularly Trichophyton rubrum , Trichophyton mentagrophytes , and Microsporum persicolor
Incubation Conditions Aerobic
Incubation Time 3 to 7 days
No. per Pack 20
Packaging Type Pack
Quantity 6 mL
Dimensions (Dia. x H) 16 x 100 mm
Height (Metric) 100 mm
Diameter (Metric) 16 mm
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.