Learn More
Medchemexpress LLC HY-P0244 1mg Medchemexpress, Dermorphin CAS:77614-16-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02441MG
Encompass_Preferred
Medchemexpress, HY-P0244 1mg Dermorphin CAS:77614-16-5 Dermorphin is a natural heptapeptide μ-opioid receptor ( MOR ) agonist found in amphibian skin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4075