Learn More
Cayman Chemical DermorphInacetate 10mg
SDP
Catalog No. 502992582
Encompass_Preferred
An opioid peptide; binds to μ-opioid receptors (Ki = 0.54 nM) and is selective for μ- over δ-opioid receptors in radioligand binding assays (Ki = 929 nM); inhibits noxious stimuli-induced neuronal firing in the nucleus lateralis anterior and ventrobasal complex in rats at 1.9 µmol/kg; inhibits the electrical stimulation-induced C fiber response in rat dorsal horn nociceptive neurons; delays gastric emptying, inhibits gastric acid secretion, and slows colonic motility in pylorus-ligated rats from 10-120 pmol/animal
Catalog No. 50-299-2582 Supplier Cayman Chemical Supplier No. 3160710 MG
An opioid peptide; binds to μ-opioid receptors (Ki = 0.54 nM) and is selective for μ- over δ-opioid receptors in radioligand binding assays (Ki = 929 nM); inhibits noxious stimuli-induced neuronal firing in the nucleus lateralis anterior and ventrobasal complex in rats at 1.9 µmol/kg; inhibits the electrical stimulation-induced C fiber response in rat dorsal horn nociceptive neurons; delays gastric emptying, inhibits gastric acid secretion, and slows colonic motility in pylorus-ligated rats from 10-120 pmol/animal