Learn More
Sino Biological Recombinant Dermatophagoides pteronyssinus Der p 1 Protein (His Tag)
Supplier:  Sino Biological 69007A08H1250
Encompass_Preferred
A DNA sequence encoding the dermatophagoides pteronyssinus DERP1 (P08176, with mutation Cys 132 Ala and Asn 150 Glu) (Met1-Leu320) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2917