Learn More
Medchemexpress LLC HY-13631E 10mg Medchemexpress, Deruxtecan CAS:1599440-13-7 Purity:>98%
SDP
Catalog No. 502029015
Encompass_Preferred
Medchemexpress, HY-13631E 10mg Deruxtecan CAS:1599440-13-7 Deruxtecan is an ADC drug-linker conjugate composed of a derivative of DX-8951 (DXd) and amaleimide-GGFG peptide linker, used for synthesizing DS-8201 and U3-1402. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9015 Supplier Medchemexpress LLC Supplier No. HY13631E10MG
Medchemexpress, HY-13631E 10mg Deruxtecan CAS:1599440-13-7 Deruxtecan is an ADC drug-linker conjugate composed of a derivative of DX-8951 (DXd) and amaleimide-GGFG peptide linker, used for synthesizing DS-8201 and U3-1402. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.