Learn More
Medchemexpress LLC HY-13631E 5mg , Deruxtecan CAS:1599440-13-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13631E5MG
Encompass_Preferred
HY-13631E 5mg Deruxtecan CAS: 1599440-13-7 Deruxtecan is an ADC drug-linker conjugate composed of a derivative of DX-8951 (DXd) and amaleimide-GGFG peptide linker, used for synthesizing DS-8201 and U3-1402. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9000