Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deruxtecan-d5 | 00-00-0 | 98.4% | C52H51D5FN9O13 | 10MG
SDP

Catalog No. 5000261933
Encompass_Preferred

Deruxtecan-d5 is a deuterium-labeled derivative of deruxtecan supplied as an off-white to light yellow solid for use as a tracer or internal standard in quantitative analysis (NMR, GC-MS, LC-MS) and antibody-drug conjugate research. It is highly pure, readily soluble in DMSO, and should be stored cold and protected from moisture and light.

  • deuterium-labeled internal standard for quantitative assays.
  • high purity: 98.4%.
  • solid form, off-white to light yellow.
  • soluble in DMSO ≥ 100 mg/mL.
  • storage: -20°C; in solvent -80°C for 6 months, -20°C for 1 month.
  • suitable for tracer studies and ADC research.

Catalog No. 50-002-61933 Supplier Medchemexpress LLC Supplier No. HY13631ES410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deruxtecan-d5 is a deuterium-labeled derivative of deruxtecan supplied as an off-white to light yellow solid for use as a tracer or internal standard in quantitative analysis (NMR, GC-MS, LC-MS) and antibody-drug conjugate research. It is highly pure, readily soluble in DMSO, and should be stored cold and protected from moisture and light.

  • deuterium-labeled internal standard for quantitative assays.
  • high purity: 98.4%.
  • solid form, off-white to light yellow.
  • soluble in DMSO ≥ 100 mg/mL.
  • storage: -20°C; in solvent -80°C for 6 months, -20°C for 1 month.
  • suitable for tracer studies and ADC research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.