Learn More
Medchemexpress LLC Deruxtecan-d5 | 1599440-13-7 | 98.4% | 1039.08 | 1 MG
SDP
Catalog No. 5000352243
Encompass_Preferred
Deruxtecan-d5 is a deuterium-labeled version of Deruxtecan. It is an ADC (Antibody-Drug Conjugate) drug-linker conjugate composed of a DX-8951 derivative (DXd) and a maleimide-GGFG peptide linker, used for synthesizing DS-8201 and U3-1402.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-52243 Supplier Medchemexpress LLC Supplier No. HY13631ES41MG
Deruxtecan-d5 is a deuterium-labeled version of Deruxtecan. It is an ADC (Antibody-Drug Conjugate) drug-linker conjugate composed of a DX-8951 derivative (DXd) and a maleimide-GGFG peptide linker, used for synthesizing DS-8201 and U3-1402.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS