Learn More
Medchemexpress LLC Deruxtecan-d5 | 98.4% | 1039.08 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13631ES4
Encompass_Preferred
Deruxtecan-d5 is a deuterium-labeled version of Deruxtecan, an ADC (Antibody-Drug Conjugate) drug-linker conjugate. It consists of a DX-8951 derivative and a maleimide-GGFG peptide linker, and is utilized in the synthesis of DS-8201 and U3-1402. This compound is intended for research use, specifically as a tracer or an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Used for synthesizing DS-8201 and U3-1402
  • Off-white to light yellow solid
  • For research use only
  • Store at -20°C, sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-51895