Learn More
Cayman Chemical D-erythro Lysosphingomyelin (d18:1), Purity: ≥98%, CAS: 1670-26-4, 5 mg
SDP
Catalog No. 502887676
Encompass_Preferred
A bioactive sphingolipid; agonist of S1P1, S1P2, and S1P3 (EC50s = 167.7, 368.1, and 482.6 nM, respectively, for the human receptors); agonist of ORG1 that induces calcium accumulation in cells overexpressing OGR1 (EC50 = ~35 nM); levels are increased in skin isolated from patients with atopic dermatitis, as well as postmortem brain from patients with Niemann-Pick disease type A
Catalog No. 50-288-7676 Supplier Cayman Chemical Supplier No. 100079475 MG
A bioactive sphingolipid; agonist of S1P1, S1P2, and S1P3 (EC50s = 167.7, 368.1, and 482.6 nM, respectively, for the human receptors); agonist of ORG1 that induces calcium accumulation in cells overexpressing OGR1 (EC50 = ~35 nM); levels are increased in skin isolated from patients with atopic dermatitis, as well as postmortem brain from patients with Niemann-Pick disease type A