Learn More
MOLECULAR DEPOT LLC Des-Acyl Ghrelin Human -100 ug
Catalog No. 5000013594
Encompass_Preferred
Des-Acyl Ghrelin (Human) is a synthetic peptide consisting of the sequence Gly-Ser-Ser-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg The molecular weight of this peptide is 3244 4 g/mol and the CAS number is 343268-91-7 This product is for in vitro research use only This product is not for human or animal use
Catalog No. 50-000-13594 Supplier MOLECULAR DEPOT LLC Supplier No. MDP0797
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Des-Acyl Ghrelin (Human) is a synthetic peptide consisting of the sequence Gly-Ser-Ser-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg The molecular weight of this peptide is 3244 4 g/mol and the CAS number is 343268-91-7 This product is for in vitro research use only This product is not for human or animal use