Learn More
Aviva Systems Biology Corp DES Antibody - middle region (ARP41483_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41483_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DES. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DES. Alternate names: CMD1I, CSM1, CSM2, FLJ12025, FLJ39719, FLJ41013, FLJ41793ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57877