Learn More
Cayman Chemical (Des-Arg9)-Bradykinin, Purity: ≥98%, CAS: 15958-92-6, 50 mg
SDP
Catalog No. 502989295
Encompass_Preferred
An active metabolite of bradykinin and a peptide agonist of the bradykinin B1 receptor; binds to bradykinin B1 and B2 receptors (IC50 = 8.1 µM in CHO cells expressing the human receptor); induces relaxation of precontracted isolated endothelium-denuded dog renal artery strips (pD2 = 8.6); decreases blood pressure in LPS-treated rabbits in a dose-dependent manner
Catalog No. 50-298-9295 Supplier Cayman Chemical Supplier No. 3113650 MG
An active metabolite of bradykinin and a peptide agonist of the bradykinin B1 receptor; binds to bradykinin B1 and B2 receptors (IC50 = 8.1 µM in CHO cells expressing the human receptor); induces relaxation of precontracted isolated endothelium-denuded dog renal artery strips (pD2 = 8.6); decreases blood pressure in LPS-treated rabbits in a dose-dependent manner