Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[Des-Arg9]-Bradykinin, MP Biomedicals™

Catalog No. ICN15279205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15279205 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15279205 Supplier MP Biomedicals, Inc Supplier No. 0215279205
Add to Cart
Add to Cart

A potent antagonist of bradykinin. Soluble in water (10mg/mL-clear colorless solution)

TRUSTED_SUSTAINABILITY

Specifications

CAS 15958-92-6
Color White
Format Lyophilized Powder
Molecular Weight (g/mol) 904.04
Product Type [Des-Arg9]-Bradykinin
Quantity 5mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.