Learn More
Bachem H-Glu(Cys-OEt)-OH
SDP
Catalog No. 50259442
Encompass_Preferred
(Des-Gly)-Glutathione-monoethyl ester (reduced) , H-Glu(Cys-OEt)-OH, 114627-30-4, C10H18N2O5S, Mr 278.33, 250mg. g-GCE, GCEE. g-GCE has been shown to be much more easily transported into isolated rat hepatocytes than glutathione. Within the cells it could be converted into reduced glutathione. Therefore it could - better than glutathione - prevent immunological cell injury in isolated rat hepatocytes treated with anti-rat liver plasma membrane rabbit antiserum. g-GCE may help to reduce oxidative stress in various biological systems by promoting the biosynthesis of glutathion. For this reason, administration of GCEE and N-acetyl-cysteine (NAC) was discussed as a potential therapeutic approach for Alzheimer disease.
Catalog No. 50-259-442 Supplier Bachem Supplier No. 4026208.0250
(Des-Gly)-Glutathione-monoethyl ester (reduced) , H-Glu(Cys-OEt)-OH, 114627-30-4, C10H18N2O5S, Mr 278.33, 250mg. g-GCE, GCEE. g-GCE has been shown to be much more easily transported into isolated rat hepatocytes than glutathione. Within the cells it could be converted into reduced glutathione. Therefore it could - better than glutathione - prevent immunological cell injury in isolated rat hepatocytes treated with anti-rat liver plasma membrane rabbit antiserum. g-GCE may help to reduce oxidative stress in various biological systems by promoting the biosynthesis of glutathion. For this reason, administration of GCEE and N-acetyl-cysteine (NAC) was discussed as a potential therapeutic approach for Alzheimer disease.