Learn More
Bachem (Des-Gly10,D-Ala6,Pro-NHEt9)-LHRH Acetate
SDP
Catalog No. 50259802
Encompass_Preferred
(Des-Gly10,D-Ala6,Pro-NHEt9)-LHRH Acetate, Pyr-His-Trp-Ser-Tyr-D-Ala-Leu-Arg-Pro-NHEt, 148029-26-9, C56H78N16O12, Mr 1167.34, 5mg. Dalarelin, Alarelin, Surfagon, AY 25205, [DAla6, Des-Gly10] LH-RH, Ethyl Amide. Potent LHRH analog in rats and mice. Alarelin reversibly delays sexual maturation in rats.Applied as single drug or in combination with a dopamin receptor antagonist as domperidone for the stimulation and synchronization of ovulation and spawning in fish farming. CAS Number (net): 79561-22-1.
Catalog No. 50-259-802 Supplier Bachem Supplier No. 4012028.0005
Add to Cart
Add to Cart
(Des-Gly10,D-Ala6,Pro-NHEt9)-LHRH Acetate, Pyr-His-Trp-Ser-Tyr-D-Ala-Leu-Arg-Pro-NHEt, 148029-26-9, C56H78N16O12, Mr 1167.34, 5mg. Dalarelin, Alarelin, Surfagon, AY 25205, [DAla6, Des-Gly10] LH-RH, Ethyl Amide. Potent LHRH analog in rats and mice. Alarelin reversibly delays sexual maturation in rats.Applied as single drug or in combination with a dopamin receptor antagonist as domperidone for the stimulation and synchronization of ovulation and spawning in fish farming. CAS Number (net): 79561-22-1.