Learn More
Bachem (Des-Gly10,D-Leu6,Pro-NHEt9)-LHRH Acetate
SDP
Catalog No. 50259799
Encompass_Preferred
(Des-Gly10,D-Leu6,Pro-NHEt9)-LHRH Acetate, Pyr-His-Trp-Ser-Tyr-D-Leu-Leu-Arg-Pro-NHEt, 74381-53-6, C59H84N16O12, Mr 1209.42, 5mg. Leuprorelin, Leuprolide, [Des-Gly10, DLeu6, Pro9] LH-RH, Ethyl Amide. Leuprolide, a highly active LHRH (GnRH) agonist stimulates the pituitary release of luteinizing hormone, causing an increase in testicular androgen production. Thus, leuprolide represents a safer alternative to estrogens, and may be preferred in the treatment of men with advanced prostate cancer. CAS Number (net): 53714-56-0.
Catalog No. 50-259-799 Supplier Bachem Supplier No. 4033014.0005
(Des-Gly10,D-Leu6,Pro-NHEt9)-LHRH Acetate, Pyr-His-Trp-Ser-Tyr-D-Leu-Leu-Arg-Pro-NHEt, 74381-53-6, C59H84N16O12, Mr 1209.42, 5mg. Leuprorelin, Leuprolide, [Des-Gly10, DLeu6, Pro9] LH-RH, Ethyl Amide. Leuprolide, a highly active LHRH (GnRH) agonist stimulates the pituitary release of luteinizing hormone, causing an increase in testicular androgen production. Thus, leuprolide represents a safer alternative to estrogens, and may be preferred in the treatment of men with advanced prostate cancer. CAS Number (net): 53714-56-0.