Learn More
Bachem (Des-Gly10,D-Ser(tBu)6,Pro-NHEt9)-LHRH Acetate
SDP
Supplier:  Bachem 4028150.0025
Encompass_Preferred
(Des-Gly10,D-Ser(tBu)6,Pro-NHEt9)-LHRH Acetate, Pyr-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-NHEt, 57982-77-1, C60H86N16O13, Mr 1239.44, 25mg. Buserelin, [DSer(tBu)6, Des-Gly10] LH-RH, Ethyl Amide. Buserelin is a longer acting synthetic peptide analog of the naturally occurring gonadotropin-releasing hormone (GnRH/LHRH). Substitution of glycine in position 6 by D-serine and that of glycinamide in position 10 by ethylamide, leads to a nonapeptide with a significantly enhanced LHRH effect. The effects of buserelin on follicle-stimulating hormone (FSH) and luteinizing hormone (LH) release are approximately 20 to 170 times higher than those of natural LHRH. CAS Number (buserelin acetate): 68630-75-1.
Catalog No. 50-382-915