Learn More
Bachem (Des-Gly10,D-Trp6,Pro-NHEt9)-LHRH High acetate salt
SDP
Catalog No. 50382891
Encompass_Preferred
(Des-Gly10,D-Trp6,Pro-NHEt9)-LHRH High acetate salt , Pyr-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-NHEt High acetate salt, 57773-65-6, C64H83N17O12, Mr 1282.47, 25mg. Deslorelin High acetate salt, [Des-Gly10, DTrp6, Pro-NHEt9] LH-RH. Short-term administration of deslorelin stimulates the formation of LH and FSH, which induce an increase in the production of testosterone and estradiol. Prolonged administration of the GnRH agonist causes a sustained down-regulation of circulating LH and FSH levels and hence suppression of steroidogenesis.
Catalog No. 50-382-891 Supplier Bachem Supplier No. 4066809.0025
Add to Cart
Add to Cart
(Des-Gly10,D-Trp6,Pro-NHEt9)-LHRH High acetate salt , Pyr-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-NHEt High acetate salt, 57773-65-6, C64H83N17O12, Mr 1282.47, 25mg. Deslorelin High acetate salt, [Des-Gly10, DTrp6, Pro-NHEt9] LH-RH. Short-term administration of deslorelin stimulates the formation of LH and FSH, which induce an increase in the production of testosterone and estradiol. Prolonged administration of the GnRH agonist causes a sustained down-regulation of circulating LH and FSH levels and hence suppression of steroidogenesis.