Learn More
CPC Scientific Glp-His-Trp-Ser-Tyr-DLeu-Leu-Arg-Pro-NHEt (acetate salt) 1MG
SDP
Supplier:  CPC Scientific LHRH006A
Encompass
Highly active LHRH agonist that stimulates the pituitary release of luteinizing hormone and increases testicular androgen, therefore serving as a safer alternative to estrogens. ONE-LETTER SEQUENCE: Glp-HWSY-DLeu-LRP-NHEtMOLECULAR FORMULA: C59H84N16O12MOLECULAR WEIGHT:1209.42STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [74381-53-6], SYNONYMS: [Des-Gly10, DLeu6, Pro9] LH-RH, Ethyl Amide, Leuprorelin, LeuprolideRESEARCH AREA: Hormonal
Catalog No. 50-210-9652