Learn More
CPC Scientific H-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Glu-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific EXED001A
Encompass
ONE-LETTER SEQUENCE: GEGTFTSELSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2MOLECULAR FORMULA: C179H277N47O59S1MOLECULAR WEIGHT:4063.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Diabetes
Catalog No. 50-210-9261