Learn More
Medchemexpress LLC [Des-His1,Glu9]-Glucagon amide TFA | 110084-95-2 | MFCD00080353 | 98.36% | 3472.67 | 10 MG
SDP
Catalog No. 5000423186
Encompass_Preferred
[Des-His1,Glu9]-Glucagon amide TFA is a potent peptide antagonist of the glucagon receptor, with a pA2 of 7.2. It is potentially useful in the study of the pathogenesis of diabetes.
  • Potent and peptide antagonist of the glucagon receptor
  • pA2 of 7.2
  • Potentially useful in the study of the pathogenesis of diabetes
Catalog No. 50-004-23186 Supplier Medchemexpress LLC Supplier No. HYP1143A10MG
[Des-His1,Glu9]-Glucagon amide TFA is a potent peptide antagonist of the glucagon receptor, with a pA2 of 7.2. It is potentially useful in the study of the pathogenesis of diabetes.
  • Potent and peptide antagonist of the glucagon receptor
  • pA2 of 7.2
  • Potentially useful in the study of the pathogenesis of diabetes