Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [Des-His1,Glu9]-Glucagon amide TFA | 98.4% | 3472.67 | 1 MG
SDP

Catalog No. 5000423601
Encompass_Preferred

[Des-His1,Glu9]-Glucagon amide TFA is a potent peptide antagonist of the glucagon receptor, with a pA2 of 7.2. It is potentially useful in the study of the pathogenesis of diabetes. This product is for research use only.

  • Potent peptide antagonist of the glucagon receptor
  • pA2 of 7.2
  • Useful in the study of diabetes pathogenesis
  • Purity of 98.4%
  • Molecular weight of 3472.67
  • Solid appearance, white to off-white color
  • Soluble in H2O at 6.67 mg/mL (1.92 mM)
  • Long-term storage for powder at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-23601 Supplier Medchemexpress LLC Supplier No. HYP1143A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[Des-His1,Glu9]-Glucagon amide TFA is a potent peptide antagonist of the glucagon receptor, with a pA2 of 7.2. It is potentially useful in the study of the pathogenesis of diabetes. This product is for research use only.

  • Potent peptide antagonist of the glucagon receptor
  • pA2 of 7.2
  • Useful in the study of diabetes pathogenesis
  • Purity of 98.4%
  • Molecular weight of 3472.67
  • Solid appearance, white to off-white color
  • Soluble in H2O at 6.67 mg/mL (1.92 mM)
  • Long-term storage for powder at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.