Learn More
CPC Scientific H-DPen-Gly-Phe-Pen-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ENKP004A
Encompass
SEQUENCE: H-DPen-Gly-Phe-Pen-OH (trifluoroacetate salt)(Pen1 and Pen4 bridge)ONE-LETTER SEQUENCE: DPen-GF-Pen(Pen&Pen bridge)MOLECULAR FORMULA: C21H30N4O5S2MOLECULAR WEIGHT: 482.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: PainREFERENCES:SKU(s): ENKP-004A, ENKP-004B
Catalog No. 50-210-9254