Learn More
Cayman Chemical Desacetyl Bisacodyl, Purity: ≥98%, CAS: 603-41-8, 50 mg
SDP
Catalog No. 502983004
Encompass_Preferred
An active metabolite of two stimulant laxatives, bisacodyl and sodium picosulfate
Catalog No. 50-298-3004 Supplier Cayman Chemical Supplier No. 2092850 MG
An active metabolite of two stimulant laxatives, bisacodyl and sodium picosulfate