Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desacetyl bisacodyl | 603-41-8 | MFCD00023496 | 99.0% | C18H15NO2 | 10MG
SDP

Catalog No. 5000260834
Encompass_Preferred

Desacetyl bisacodyl is the active metabolite of the laxative bisacodyl, provided as a high-purity white to off-white solid for laboratory research. It has formula C18H15NO2 and a molecular weight of 277.32 g/mol. The compound is offered in small milligram pack sizes and includes guidance for cold-chain storage to preserve stability.

  • Active metabolite of bisacodyl.
  • High purity (99.0%).
  • White to off-white solid physical form.
  • Molecular formula C18H15NO2; MW 277.32 g/mol.
  • Available in small milligram pack sizes for research use.
  • Storage: 4°C under nitrogen; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-60834 Supplier Medchemexpress LLC Supplier No. HY13163710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Desacetyl bisacodyl is the active metabolite of the laxative bisacodyl, provided as a high-purity white to off-white solid for laboratory research. It has formula C18H15NO2 and a molecular weight of 277.32 g/mol. The compound is offered in small milligram pack sizes and includes guidance for cold-chain storage to preserve stability.

  • Active metabolite of bisacodyl.
  • High purity (99.0%).
  • White to off-white solid physical form.
  • Molecular formula C18H15NO2; MW 277.32 g/mol.
  • Available in small milligram pack sizes for research use.
  • Storage: 4°C under nitrogen; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.