Learn More
Cayman Chemical Desacetyl Cefapirin (sodium salt), Purity: ≥95%, CAS: 104557-24-6, 25 mg
SDP
Catalog No. 502940093
Encompass_Preferred
An active metabolite of cefapirin; active against E. coli, K. pneumoniae, P. mirabilis, and S. aureus (MICs = 120, 24, 34, and 0.42 µg/ml, respectively)
Catalog No. 50-294-0093 Supplier Cayman Chemical Supplier No. 2757825 MG
An active metabolite of cefapirin; active against E. coli, K. pneumoniae, P. mirabilis, and S. aureus (MICs = 120, 24, 34, and 0.42 µg/ml, respectively)