Learn More
Medchemexpress LLC Desacetylcinobufagin | 4026-95-3 | 400.51 | 1 MG
SDP
Catalog No. 5000416917
Encompass_Preferred
Desacetylcinobufagin is a natural compound primarily used for microbial transformation research. This product is intended for research use only.
  • Derived from animals (toad)
  • Exhibits cytotoxicity against HL-60 cells (IC50 of 1 μg/mL)
  • Exhibits cytotoxicity against KB cells (IC50 of 0.44 μg/mL)
  • Used in microbial transformation studies
Catalog No. 50-004-16917 Supplier Medchemexpress LLC Supplier No. HYN08811MG
Desacetylcinobufagin is a natural compound primarily used for microbial transformation research. This product is intended for research use only.
  • Derived from animals (toad)
  • Exhibits cytotoxicity against HL-60 cells (IC50 of 1 μg/mL)
  • Exhibits cytotoxicity against KB cells (IC50 of 0.44 μg/mL)
  • Used in microbial transformation studies