Learn More
Medchemexpress LLC Desacetylcinobufotalin | 4099-30-3 | 416.51 | 1 MG
SDP
Catalog No. 5000416118
Encompass_Preferred
Desacetylcinobufotalin is a natural compound and an apoptosis inducer. It demonstrates a marked inhibitory effect on HepG2 cells, with an IC50 value of 0.0279 μmol/mL. It is for research use only and not sold to patients. The product appears as a solid, white to light yellow in color.
  • Purity: 99.41%
  • Induces apoptosis
  • Inhibitory effect on HepG2 cells with anIC50 of 0.0279 μmol/mL
  • Cytotoxicity against human HL60 cells: IC50 4.3 μg/mL
  • Cytotoxicity against human KB cells: IC50 10 μg/mL
Catalog No. 50-004-16118 Supplier Medchemexpress LLC Supplier No. HYN08821MG
Desacetylcinobufotalin is a natural compound and an apoptosis inducer. It demonstrates a marked inhibitory effect on HepG2 cells, with an IC50 value of 0.0279 μmol/mL. It is for research use only and not sold to patients. The product appears as a solid, white to light yellow in color.
  • Purity: 99.41%
  • Induces apoptosis
  • Inhibitory effect on HepG2 cells with anIC50 of 0.0279 μmol/mL
  • Cytotoxicity against human HL60 cells: IC50 4.3 μg/mL
  • Cytotoxicity against human KB cells: IC50 10 μg/mL