Learn More
AdipoGen Desacetylcolchicine tartrate
SDP
Catalog No. 502561155
Encompass_Preferred
Chemical. CAS 49720-72-1. Formula C20H23NO5 . C4H6O6. MW 357.4 . 150.1. Synthetic. Anticancer colchicine analog. Microtubule polymerization inhibitor. Prevents cell division.
Catalog No. 50-256-1155 Supplier AdipoGen Supplier No. CDXD0270M025
Chemical. CAS 49720-72-1. Formula C20H23NO5 . C4H6O6. MW 357.4 . 150.1. Synthetic. Anticancer colchicine analog. Microtubule polymerization inhibitor. Prevents cell division.