Learn More
Medchemexpress LLC HY-W015346 100mg Medchemexpress, Desaminotyrosine CAS:501-97-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW015346100MG
Encompass_Preferred
Medchemexpress, HY-W015346 100mg Desaminotyrosine CAS:501-97-3 Desaminotyrosine is a microbially associated metabolite protecting from influenza through augmentation of type I interferon signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4601