Learn More
Medchemexpress LLC Desaminotyrosine | 501-97-3 | 99.9% | 166.18 g/mol | C9H10O3 | 25 G
SDP
Catalog No. 5000303015
Encompass_Preferred
Desaminotyrosine (3-(4-hydroxyphenyl)propionic acid) is a purified analytical standard supplied as a solid for research and analytical applications. It is reported to be a microbially associated metabolite that augments type I interferon signaling and has been used in influenza-related research. The material is intended for qualitative and quantitative assays and is compatible with common analytical techniques.
Catalog No. 50-003-03015 Supplier Medchemexpress LLC Supplier No. 25GHYW015346
Desaminotyrosine (3-(4-hydroxyphenyl)propionic acid) is a purified analytical standard supplied as a solid for research and analytical applications. It is reported to be a microbially associated metabolite that augments type I interferon signaling and has been used in influenza-related research. The material is intended for qualitative and quantitative assays and is compatible with common analytical techniques.